α-Bromo-2-chloro-4-fluorotoluene 98%
Supplier: Apollo Scientific
Danger
Synonyms: 1-(Bromomethyl)-2-chloro-4-fluorobenzene, 2-Chloro-4-fluorobenzyl bromide
PC0070-100G PC0070-10G PC0070-25G
APOSPC0070-100GEA 72.5 EUR
APOSPC0070-100G APOSPC0070-10G APOSPC0070-25G
α-Bromo-2-chloro-4-fluorotoluene 98%
α-Bromo-2-chloro-4-fluorotoluene
Formula: C₇H₅BrClF
Boiling Pt: 97…99 °C/10mmHg
Melting Pt: 33…39 °C
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00236025
CAS Number: 45767-66-6
UN: 3261
ADR: 8,II