3,5-Difluoro-o-phenylenediamine
Supplier: Apollo Scientific
Warning
Synonyms: 3,5-Difluoro-1,2-phenylenediamine, 1,2-Diamino-3,5-difluorobenzene, 3,5-Difluorobenzene-1,2-diamine
PC0143-1G PC0143-25G PC0143-5G
APOSPC0143-1GEA 21.9 EUR
APOSPC0143-1G APOSPC0143-25G APOSPC0143-5G
3,5-Difluoro-o-phenylenediamine
3,5-Difluoro-o-phenylenediamine
Formula: C₆H₆F₂N₂
MW: 144.12 g/mol
Melting Pt: 48.5…49.5 °C
Flash Pt: 108 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00973899
CAS Number: 2369-29-1