3-Chloro-2-fluoro-6-(trifluoromethyl)benzoic acid 98%
Supplier: Apollo Scientific
Warning
PC0151-25G PC0151-5G PC0151-1G
APOSPC0151-25GEA 281 EUR
APOSPC0151-25G APOSPC0151-5G APOSPC0151-1G
3-Chloro-2-fluoro-6-(trifluoromethyl)benzoic acid 98%
3-Chloro-2-fluoro-6-(trifluoromethyl)benzoic acid
Formula: C₈H₃ClF₄O₂
MW: 242.56 g/mol
Melting Pt: 96…98 °C
Storage Temperature: Ambient
MDL Number: MFCD01631342
CAS Number: 186517-41-9