(3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl)acetic acid 98%
Supplier: Apollo Scientific
Warning
PC0166-1G PC0166-5G PC0166-10G
APOSPC0166-1GEA 78.3 EUR
APOSPC0166-1G APOSPC0166-5G APOSPC0166-10G
(3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl)acetic acid 98%
(3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl)acetic acid
Formula: C₉H₅ClF₄O₂
Melting Pt: 134…136 °C
Storage Temperature: Ambient
MDL Number: MFCD01631516
CAS Number: 261763-13-7