3-Chloro-4-(trifluoromethoxy)aniline 98%
Supplier: Apollo Scientific
Warning
PC0170-5G PC0170-100G PC0170-25G
APOSPC0170-5GEA 21.9 EUR
APOSPC0170-5G APOSPC0170-100G APOSPC0170-25G
3-Chloro-4-(trifluoromethoxy)aniline 98%
3-Chloro-4-(trifluoromethoxy)aniline
Formula: C₇H₅ClF₃NO
MW: 211.57 g/mol
Boiling Pt: 241
Melting Pt: 29…30 °C
Density: 1.454 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00190126
CAS Number: 64628-73-5