4-Carboxy-2-chloro-a,a,a-trifluoroanisole 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Chloro-4-(trifluoromethoxy)benzoic acid
PC0177-100G PC0177-25G PC0177-5G PC0177-1G PC0177-50G
APOSPC0177-100GEA 473 EUR
APOSPC0177-100G APOSPC0177-25G APOSPC0177-5G APOSPC0177-1G APOSPC0177-50G
4-Carboxy-2-chloro-a,a,a-trifluoroanisole 97%
4-Carboxy-2-chloro-a,a,a-trifluoroanisole
Formula: C₈H₄ClF₃O₃
MW: 240.56 g/mol
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD01631556
CAS Number: 158580-93-9