5-Chloro-2-(trifluoromethyl)phenylacetonitrile 98%
Supplier: Apollo Scientific
PC0204-10G PC0204-5G PC0204-1G
APOSPC0204-10GEA 405 EUR
APOSPC0204-10G APOSPC0204-5G APOSPC0204-1G
5-Chloro-2-(trifluoromethyl)phenylacetonitrile 98%
5-Chloro-2-(trifluoromethyl)phenylacetonitrile
Formula: C₉H₅ClF₃N
MW: 219.6 g/mol
Melting Pt: 61…63 °C
Storage Temperature: Ambient
MDL Number: MFCD01631357
CAS Number: 261763-26-2