(2,3-Difluoro-4-methylphenyl)acetonitrile
Supplier: Apollo Scientific
Warning
PC0263-1G PC0263-5G PC0263-10G
APOSPC0263-1GEA 50.9 EUR
APOSPC0263-1G APOSPC0263-5G APOSPC0263-10G
(2,3-Difluoro-4-methylphenyl)acetonitrile
(2,3-Difluoro-4-methylphenyl)acetonitrile
Formula: C₉H₇F₂N
Storage Temperature: Ambient
MDL Number: MFCD01631648
CAS Number: 261944-01-8
UN: 3439
ADR: 6.1,III