2-Fluorobenzenesulphonamide 97%
Supplier: Apollo Scientific
Danger
PC0267-1G PC0267-25G PC0267-5G
APOSPC0267-1GEA 14.7 EUR
APOSPC0267-1G APOSPC0267-25G APOSPC0267-5G
2-Fluorobenzenesulphonamide 97%
2-Fluorobenzenesulphonamide
Formula: C₆H₆FNO₂S
MW: 175.18 g/mol
Melting Pt: 162…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00042283
CAS Number: 30058-40-3
UN: 2811
ADR: 6.1 III