3-Fluoro-4-methylbenzaldehyde 96%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Fluoro-4-tolualdehyde, 3-Fluoro-p-tolylaldehyde, 3-Fluoro-p-tolualdehyde
PC0339-100G PC0339-25G
APOSPC0339-100GEA 219 EUR
APOSPC0339-100G APOSPC0339-25G
3-Fluoro-4-methylbenzaldehyde 96%
3-Fluoro-4-methylbenzaldehyde
Formula: C₈H₇FO
MW: 138.14 g/mol
Boiling Pt: 206 °C
Density: 1.133 g/cm³ (25 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD01631532
CAS Number: 177756-62-6