3-Fluoro-2-(trifluoromethyl)benzamide
Supplier: Apollo Scientific
Warning
PC0386-1G PC0386-5G PC0386-25G
APOSPC0386-1GEA 128 EUR
APOSPC0386-1G APOSPC0386-5G APOSPC0386-25G
3-Fluoro-2-(trifluoromethyl)benzamide
3-Fluoro-2-(trifluoromethyl)benzamide
Formula: C₈H₅F₄NO
Storage Temperature: Ambient
MDL Number: MFCD01631492
CAS Number: 287398-80-5