2-Methyl-4-(trifluoromethoxy)benzoic acid 97%
Supplier: Apollo Scientific
Warning
PC0416-1G PC0416-5G
APOSPC0416-1GEA 71.2 EUR
APOSPC0416-1G APOSPC0416-5G
2-Methyl-4-(trifluoromethoxy)benzoic acid 97%
2-Methyl-4-(trifluoromethoxy)benzoic acid
Formula: C₉H₇F₃O₃
MW: 220.15 g/mol
Melting Pt: 88…89 °C
Storage Temperature: Ambient
MDL Number: MFCD01631542
CAS Number: 261951-91-1