3,4,5-Trifluoroanisole
Supplier: Apollo Scientific
Warning
Synonyms: 1,2,3-Trifluoro-5-methoxybenzene, Trifluoro-3,4,5-anisole, 1-Methoxy-3,4,5-trifluorobenzene
PC0513-10G PC0513-5G PC0513-25G
APOSPC0513-10GEA 127 EUR
APOSPC0513-10G APOSPC0513-5G APOSPC0513-25G
3,4,5-Trifluoroanisole
3,4,5-Trifluoroanisole
Formula: C₇H₅F₃O
Boiling Pt: 39 °C/1mmHg
Flash Pt: 52 °C
Storage Temperature: Ambient
MDL Number: MFCD01631451
CAS Number: 203245-17-4
UN: 3271
ADR: 3,III