1-(3-Chloro-5-(trifluoromethyl)-2-pyridinyl)tetrahydro-4(1H)-pyridinone
Supplier: Apollo Scientific
Warning
Synonyms: 1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]piperidin-4-one
PC0530-1G PC0530-5G
APOSPC0530-1GEA 147 EUR
APOSPC0530-1G APOSPC0530-5G
1-(3-Chloro-5-(trifluoromethyl)-2-pyridinyl)tetrahydro-4(1H)-pyridinone
1-(3-Chloro-5-(trifluoromethyl)-2-pyridinyl)tetrahydro-4(1H)-pyridinone
Formula: C₁₁H₁₀ClF₃N₂O
Melting Pt: 62…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00231641
CAS Number: 339029-35-5