1,3-Difluoroacetone 98%
Supplier: Apollo Scientific
Danger
PC066-5G PC066-1G
APOSPC066-5GEA 331 EUR
APOSPC066-5G APOSPC066-1G
1,3-Difluoroacetone 98%
1,3-Difluoroacetone
Formula: C₃H₄F₂O
Boiling Pt: 101…102 °C
Density: 1.307 g/cm³ (25 °C)
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD07368665
CAS Number: 453-14-5
UN: 2929
ADR: 6.1+3,II