4'-(2-Fluorophenyl)propiophenone 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-(2'-Fluorobiphenyl-4-yl)propan-1-one
PC0701-10G
PC0701-1G
APOSPC0701-10GEA
168
EUR
APOSPC0701-10G
APOSPC0701-1G
4'-(2-Fluorophenyl)propiophenone 97%
4'-(2-Fluorophenyl)propiophenone
Formula:
C₁₅H₁₃FO Melting Pt: 76…77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236612 CAS Number: 37989-92-7 |