2-Chloro-5-fluorobenzaldehyde 95%
Supplier: Apollo Scientific
Warning
PC0777-25G PC0777-5G
APOSPC0777-25GEA 56.7 EUR
APOSPC0777-25G APOSPC0777-5G
2-Chloro-5-fluorobenzaldehyde 95%
2-Chloro-5-fluorobenzaldehyde
Formula: C₇H₄ClFO
Melting Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD03788511
CAS Number: 84194-30-9