2-Hydroxy-3-(trifluoromethyl)benzaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 3-(Trifluoromethyl)salicylaldehyde
PC0841-1G PC0841-250MG PC0841-500MG
APOSPC0841-1GEA 107 EUR
APOSPC0841-1G APOSPC0841-250MG APOSPC0841-500MG
2-Hydroxy-3-(trifluoromethyl)benzaldehyde
2-Hydroxy-3-(trifluoromethyl)benzaldehyde
Formula: C₈H₅F₃O₂
MW: 190.12 g/mol
Melting Pt: 54…59 °C
Storage Temperature: Ambient
MDL Number: MFCD03788529
CAS Number: 336628-67-2