2H-Perfluoro-5,8-dimethyl-3,6,9-trioxadodecane 95%
Supplier: Apollo Scientific
Warning
PC0919-5G PC0919-1G PC0919-25G
APOSPC0919-5GEA 147 EUR
APOSPC0919-5G APOSPC0919-1G APOSPC0919-25G
2H-Perfluoro-5,8-dimethyl-3,6,9-trioxadodecane 95%
2H-Perfluoro-5,8-dimethyl-3,6,9-trioxadodecane
Formula: C₁₁HF₂₃O₃
Boiling Pt: 151…154 °C
Melting Pt: –106.7 °C
Density: 1.723 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00054715
CAS Number: 3330-16-3