Diethyl (2,4,6-trifluorophenyl)malonate
Supplier: Apollo Scientific
Warning
Synonyms: Diethyl 2-(2,4,6-trifluorophenyl)malonate
PC10101-1G PC10101-250MG
APOSPC10101-1GEA 75.4 EUR
APOSPC10101-1G APOSPC10101-250MG
Diethyl (2,4,6-trifluorophenyl)malonate
Diethyl (2,4,6-trifluorophenyl)malonate
Formula: C₁₃H₁₃F₃O₄
MW: 290.2351 g/mol
Melting Pt: 46…49 °C
Storage Temperature: Ambient
MDL Number: MFCD04039210
CAS Number: 262609-07-4