7-Chloro-2,4-bis(trifluoromethyl)[1,8]naphthyridine
Supplier: Apollo Scientific
PC10110-1G PC10110-500MG PC10110-5G
APOSPC10110-1GEA 533 EUR
APOSPC10110-1G APOSPC10110-500MG APOSPC10110-5G
7-Chloro-2,4-bis(trifluoromethyl)[1,8]naphthyridine
7-Chloro-2,4-bis(trifluoromethyl)[1,8]naphthyridine
Formula: C₁₀H₃ClF₆N₂
MW: 300.59 g/mol
Melting Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00140156
CAS Number: 106582-41-6