1,3-Dimethyl-5-[3-(trifluoromethyl)phenoxy]-1H-pyrazole-4-carbaldehyde
Supplier: Apollo Scientific
PC10131-500MG PC10131-1G
APOSPC10131-500MGEA 496 EUR
APOSPC10131-500MG APOSPC10131-1G
1,3-Dimethyl-5-[3-(trifluoromethyl)phenoxy]-1H-pyrazole-4-carbaldehyde
1,3-Dimethyl-5-[3-(trifluoromethyl)phenoxy]-1H-pyrazole-4-carbaldehyde
Formula: C₁₃H₁₁F₃N₂O₂
Melting Pt: 84…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00215392
CAS Number: 109925-28-2