Ethyl-8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
PC10143-1G PC10143-5G
APOSPC10143-1GEA 29 EUR
APOSPC10143-1G APOSPC10143-5G
Ethyl-8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Ethyl 8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Formula: C₁₁H₈ClF₃N₂O₂
MW: 292.65 g/mol
Melting Pt: 132…134 °C
Storage Temperature: Ambient
MDL Number: MFCD01833013
CAS Number: 353258-31-8