4,4'-Di(hydroxy)octafluorobiphenyl hydrate
Supplier: Apollo Scientific
Warning
Synonyms: 2,2',3,3',5,5',6,6'-Octafluoro-4,4'-biphenol monohydrate
PC1015-250MG PC1015-500MG
APOSPC1015-250MGEA 21.9 EUR
APOSPC1015-250MG APOSPC1015-500MG
4,4'-Di(hydroxy)octafluorobiphenyl hydrate
4,4'-Di(hydroxy)octafluorobiphenyl hydrate
Formula: C₁₂H₄F₈O₃
MW: 348.15 g/mol
Melting Pt: 209…211 °C
Storage Temperature: Ambient
MDL Number: MFCD00149466
CAS Number: 205926-99-4