1-((4-Fluorophenyl)sulphonyl)-4-pyridinecarboxylic acid
Supplier: Apollo Scientific
Warning
PC10241-1G PC10241-250MG PC10241-5G
APOSPC10241-1GEA 109 EUR
APOSPC10241-1G APOSPC10241-250MG APOSPC10241-5G
1-((4-Fluorophenyl)sulphonyl)-4-pyridinecarboxylic acid
1-((4-Fluorophenyl)sulphonyl)-4-pyridinecarboxylic acid
Formula: C₁₂H₁₄FNO₄S
Melting Pt: 174…176 °C
Storage Temperature: Ambient
MDL Number: MFCD02331439
CAS Number: 326907-70-4