2-[(3,4,4-Trifluoro-3-butenyl)sulphanyl]nicotinic acid
Supplier: Apollo Scientific
Synonyms: 2-[(3,4,4-Trifluorobut-3-en-1-yl)thio]nicotinic acid
PC10270-1G PC10270-500MG PC10270-5G
APOSPC10270-1GEA 275 EUR
APOSPC10270-1G APOSPC10270-500MG APOSPC10270-5G
2-[(3,4,4-Trifluoro-3-butenyl)sulphanyl]nicotinic acid
2-[(3,4,4-Trifluoro-3-butenyl)sulphanyl]nicotinic acid
Formula: C₁₀H₈F₃NO₂S
Melting Pt: 123…125 °C
Storage Temperature: Ambient
MDL Number: MFCD01814935
CAS Number: 453557-66-9