Pentafluorophenyl-2,5-dichlorobenzenesulphonate
Supplier: Apollo Scientific
PC10323-1G PC10323-500MG PC10323-5G
APOSPC10323-1GEA 275 EUR
APOSPC10323-1G APOSPC10323-500MG APOSPC10323-5G
Pentafluorophenyl-2,5-dichlorobenzenesulphonate
Formula: C₁₂H₃Cl₂F₅O₃S
Melting Pt: 122…124 °C
Storage Temperature: Ambient
MDL Number: MFCD05975133
CAS Number: 885950-40-3