4-Hydroxy-2-(trifluoromethyl)benzonitrile 98%
Supplier: Apollo Scientific
Warning
PC10356-1G PC10356-5G PC10356-250MG
APOSPC10356-1GEA 143 EUR
APOSPC10356-1G APOSPC10356-5G APOSPC10356-250MG
4-Hydroxy-2-(trifluoromethyl)benzonitrile 98%
4-Hydroxy-2-(trifluoromethyl)benzonitrile
Formula: C₈H₄F₃NO
MW: 187.12 g/mol
Melting Pt: 117…124 °C
Storage Temperature: Ambient
MDL Number: MFCD06797340
CAS Number: 320-42-3