2-(4-Fluorophenyl)-5-nitropyridine
Supplier: Apollo Scientific
PC10367-1G PC10367-500MG PC10367-5G
APOSPC10367-1GEA 275 EUR
APOSPC10367-1G APOSPC10367-500MG APOSPC10367-5G
2-(4-Fluorophenyl)-5-nitropyridine
2-(4-Fluorophenyl)-5-nitropyridine
Formula: C₁₁H₇FN₂O₂
Melting Pt: 132…136 °C
Storage Temperature: Ambient
MDL Number: MFCD06200966
CAS Number: 886361-78-0