5-Chloro-2-fluoropyridine 98%
Supplier: Apollo Scientific
Danger
PC10416-25G PC10416-5G PC10416-100G
APOSPC10416-25GEA 26.1 EUR
APOSPC10416-25G APOSPC10416-5G APOSPC10416-100G
5-Chloro-2-fluoropyridine 98%
5-Chloro-2-chloropyridine
Formula: C₅H₃ClFN
Boiling Pt: 149 °C
Density: 1.311 g/cm³ (25 °C)
Flash Pt: 52 °C
Storage Temperature: Ambient
MDL Number: MFCD04112513
CAS Number: 1480-65-5
UN: 1993
ADR: 3,III