2-Bromo-2',4',6'-trifluoroacetophenone
Supplier: Apollo Scientific
Danger
Synonyms: 2,4,6-Trifluorophenacyl bromide
PC10424-100MG PC10424-1G PC10424-5G
APOSPC10424-100MGEA 21.9 EUR
APOSPC10424-100MG APOSPC10424-1G APOSPC10424-5G
2-Bromo-2',4',6'-trifluoroacetophenone
2-Bromo-2',4',6'-trifluoroacetophenone
Formula: C₈H₄BrF₃O
Melting Pt: 36…38 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368690
CAS Number: 746630-36-4
UN: 1759
ADR: 8,II