4-Chloro-2-(trifluoromethyl)aniline 99%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Amino-5-chloro-α,α,α-trifluorotoluene, 2-Amino-5-chloro-α,α,α-trifluortoluene, 2-Amino-5-chlorobenzotrifluoride, 4-Chloro-α,α,α-trifluoro-o-toluidine
PC1050-100G PC1050-25G
APOSPC1050-100GEA 24.8 EUR
APOSPC1050-100G APOSPC1050-25G
4-Chloro-2-(trifluoromethyl)aniline 99%
4-Chloro-2-(trifluoromethyl)aniline
Formula: C₇H₅ClF₃N
Boiling Pt: 66…67 °C/3mmHg
Melting Pt: 44447 °C
Density: 1.386 g/cm³ (25 °C)
Flash Pt: 95 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007841
CAS Number: 445-03-4
EINECS: 207-151-3