2-Chloro-4-(trifluoromethyl)phenylboronic acid
Supplier: Apollo Scientific
Warning
Synonyms: (2-Chloro-4-(trifluoromethyl)phenyl)metaboric acid, Dihydroxy(2-chloro-4-(trifluoromethyl)phenyl)borane, 2-Chloro-4-(trifluoromethyl)benzeneboronic acid
PC10519-25G PC10519-1G PC10519-5G
APOSPC10519-25GEA 277 EUR
APOSPC10519-25G APOSPC10519-1G APOSPC10519-5G
2-Chloro-4-(trifluoromethyl)phenylboronic acid
2-Chloro-4-(trifluoromethyl)phenylboronic acid
Formula: C₇H₅BClF₃O₂
MW: 224.37
Melting Pt: 110…112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02684337
CAS Number: 254993-59-4