5-Bromo-2-(trifluoromethyl)pyridine 97%
Supplier: Apollo Scientific
Danger
Synonyms: 5-Bromo-2-trifluoromethylpyridine
PC1054-100G PC1054-25G PC1054-5G
APOSPC1054-100GEA 267 EUR
APOSPC1054-100G APOSPC1054-25G APOSPC1054-5G
5-Bromo-2-(trifluoromethyl)pyridine 97%
5-Bromo-2-(trifluoromethyl)pyridine
Formula: C₆H₃BrF₃N
MW: 226.00 g/mol
Boiling Pt: 68…70 °C/10mmHg
Melting Pt: 39…45 °C
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD06657686
CAS Number: 436799-32-5
UN: 2811
ADR: 6.1 III