2-Fluoropropionic acid 97%
Supplier: Apollo Scientific
Danger
PC1057-1G PC1057-250MG PC1057-5G
APOSPC1057-1GEA 48 EUR
APOSPC1057-1G APOSPC1057-250MG APOSPC1057-5G
2-Fluoropropionic acid 97%
2-Fluoropropionic acid
Formula: C₃H₅FO₂
Boiling Pt: 66…67 °C/30mmHg
Density: 1.181 g/cm³ (25 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD06247714
CAS Number: 6087-13-4
UN: 3265
ADR: 8,II