2,6-Difluoro-4-methoxyphenol
Supplier: Apollo Scientific
Danger
Synonyms: 3,5-Difluor-4-hydroxyanisole
PC10709-1G PC10709-5G
APOSPC10709-1GEA 79.9 EUR
APOSPC10709-1G APOSPC10709-5G
2,6-Difluoro-4-methoxyphenol
2,6-Difluoro-4-methoxyphenol
Formula: C₇H₆F₂O₂
MW: 160.12 g/mol
Melting Pt: 67…71 °C
Storage Temperature: Ambient
MDL Number: MFCD04115926
CAS Number: 886498-93-7