4-Amino-2-(trifluoromethyl)benzonitrile 98%
Supplier: Apollo Scientific
Danger
Synonyms: 4-Cyano-3-trifluoromethylaniline
PC1070L-100G PC1070L-250G PC1070L-500G
APOSPC1070L-100GEA 29 EUR
APOSPC1070L-100G APOSPC1070L-250G APOSPC1070L-500G
4-Amino-2-(trifluoromethyl)benzonitrile 98%
4-Amino-2-(trifluoromethyl)benzonitrile
Formula: C₈H₅F₃N₂
MW: 186.14 g/mol
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00042155
CAS Number: 654-70-6
UN: 3439
ADR: 6.1 III