5-Amino-1-(4-fluorophenyl)-1H-pyrazole-4-carbonitrile 97%
Supplier: Apollo Scientific
Warning
PC1070M-1G PC1070M-5G
APOSPC1070M-1GEA 27.3 EUR
APOSPC1070M-1G APOSPC1070M-5G
5-Amino-1-(4-fluorophenyl)-1H-pyrazole-4-carbonitrile 97%
5-Amino-1-(4-fluorophenyl)-1H-pyrazole-4-carbonitrile
Formula: C₁₀H₇FN₄
Melting Pt: 164…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00102729
CAS Number: 51516-70-2