2,8-Difluoro-5-(trifluoromethyl)-5H-dibenzo[b,d]thiophen-5-ium trifluoromethanesulphonate 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2,8-Difluoro-5-(trifluoromethyl)-5H-dibenzo[b,d]thiophen-5-ium triflate, Umemoto's Reagent II
PC10719-25G PC10719-5G
APOSPC10719-25GEA 464 EUR
APOSPC10719-25G APOSPC10719-5G
2,8-Difluoro-5-(trifluoromethyl)-5H-dibenzo[b,d]thiophen-5-ium trifluoromethanesulphonate 98%
2,8-Difluoro-5-(trifluoromethyl)-5H-dibenzo[b,d]thiophen-5-ium trifluoromethanesulphonate
Formula: C₁₄H₆F₈O₃S₂
MW: 438.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30725753
CAS Number: 1961266-44-3