4-Amino-3-(trifluoromethoxy)benzoic acid 97%
Supplier: Apollo Scientific
Warning
PC1100BD-1G PC1100BD-25G PC1100BD-5G
APOSPC1100BD-1GEA 21.9 EUR
APOSPC1100BD-1G APOSPC1100BD-25G APOSPC1100BD-5G
4-Amino-3-(trifluoromethoxy)benzoic acid 97%
4-Amino-3-(trifluoromethoxy)benzoic acid
Formula: C₈H₆F₃NO₃
MW: 221.14 g/mol
Melting Pt: 168…169 °C
Storage Temperature: Ambient
MDL Number: MFCD00205143
CAS Number: 175278-22-5