2,6-Difluoro-3-methoxybenzaldehyde
Supplier: Apollo Scientific
Warning
PC1127-1G PC1127-5G
APOSPC1127-1GEA 66.7 EUR
APOSPC1127-1G APOSPC1127-5G
2,6-Difluoro-3-methoxybenzaldehyde
2,6-Difluoro-3-methoxybenzaldehyde
Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115899
CAS Number: 149949-30-4