3,6-Difluorosalicylaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 3,6-Difluoro-2-hydroxybenzaldehyde
PC1144-1G PC1144-25G PC1144-5G
APOSPC1144-1GEA 84.1 EUR
APOSPC1144-1G APOSPC1144-25G APOSPC1144-5G
3,6-Difluorosalicylaldehyde
3,6-Difluorosalicylaldehyde
Formula: C₇H₄F₂O₂
MW: 158.1023 g/mol
Melting Pt: 72…78 °C
Storage Temperature: Ambient
MDL Number: MFCD08275281
CAS Number: 502762-92-7