4,5-Difluorophthalic anhydride 95%
Supplier: Apollo Scientific
Warning
Synonyms: 4,5-Difluorophthalic Anhydride
PC1162-10G PC1162-1G PC1162-5G
APOSPC1162-10GEA 567 EUR
APOSPC1162-10G APOSPC1162-1G APOSPC1162-5G
4,5-Difluorophthalic anhydride 95%
4,5-Difluorophthalic anhydride
Formula: C₈H₂F₂O₃
MW: 184.1 g/mol
Melting Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD08695033
CAS Number: 18959-30-3