4,4'-(Hexafluoroisopropylidene)dianiline 98%
Supplier: Apollo Scientific
Warning
PC1225G-1G PC1225G-5G PC1225G-25G
APOSPC1225G-1GEA 21.9 EUR
APOSPC1225G-1G APOSPC1225G-5G APOSPC1225G-25G
4,4'-(Hexafluoroisopropylidene)dianiline 98%
4,4'-(Hexafluoroisopropylidene)dianiline
Formula: C₁₅H₁₂F₆N₂
Melting Pt: 195…202 °C
Storage Temperature: Ambient
MDL Number: MFCD00039146
CAS Number: 1095-78-9