Bis(4-fluorophenyl)ether 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4,4'-Oxybis(fluorobenzene), 4,4'-Difluorodiphenyl Ether
PC1227J-1G PC1227J-5G
APOSPC1227J-1GEA 21.9 EUR
APOSPC1227J-1G APOSPC1227J-5G
Bis(4-fluorophenyl)ether 97%
Bis(4-fluorophenyl)ether
Formula: C₁₂H₈F₂O
MW: 206.19 g/mol
Boiling Pt: 232…240 °C/743mmHg
Density: 1.23 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00013552
CAS Number: 330-93-8
EINECS: 206-358-6