Bismuth(III) fluoride 99,5%
Supplier: Apollo Scientific
Danger
PC1230G-5G PC1230G-1G
APOSPC1230G-5GEA 0 EUR
APOSPC1230G-5G APOSPC1230G-1G
Bismuth(III) fluoride 99,5%
Bismuth (III) fluoride
Formula: BiF₃
MW: 265.98 g/mol
Boiling Pt: 230
Melting Pt: 727 °C
Density: 5.32 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010893
CAS Number: 7787-61-3
UN: 3260
ADR: 8 II
Merck Index: 12,01305