1,1,1-Trifluoro-2-(trifluoromethyl)-4-penten-2-ol 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Allylhexafluoroisopropanol
PC1259-25G PC1259-5G PC1259-1G
APOSPC1259-25GEA 321 EUR
APOSPC1259-25G APOSPC1259-5G APOSPC1259-1G
1,1,1-Trifluoro-2-(trifluoromethyl)-4-penten-2-ol 98%
1,1,1-Trifluoro-2-(trifluoromethyl)-4-penten-2-ol
Formula: C₆H₆F₆O
Boiling Pt: 42.8 °C/68mmHg
Density: 1.336 g/cm³ (25 °C)
Flash Pt: 52 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03092986
CAS Number: 646-97-9
EINECS: 613-651-6
UN: 2924
ADR: 3+8,III