N,O-Bis(trimethylsilyl)trifluoroacetamide 99+%
Supplier: Apollo Scientific
Danger
Synonyms: Trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)acetimidate, Bis(Trimethylsilyl)Trifluoroacetamide, BSTFA
PC1260-100G PC1260-250G PC1260-25G
APOSPC1260-100GEA 43.5 EUR
APOSPC1260-100G APOSPC1260-250G APOSPC1260-25G
N,O-Bis(trimethylsilyl)trifluoroacetamide 99+%
N,O-Bis(trimethylsilyl)trifluoroacetamide
Formula: C₈H₁₈F₃NOSi₂
Boiling Pt: 45…50 °C/14mmHg
Melting Pt: –10 °C
Density: 0.969 g/cm³ (25 °C)
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00008269
CAS Number: 25561-30-2
UN: 2924
ADR: 3+8,III