Perfluoro-2,5,8,11,14,17,20-heptamethyl-3,6,9,12,15,18,21-heptaoxatetracosanoyl fluoride 90%
Supplier: Apollo Scientific
Danger
PC1295-25G PC1295-5G
APOSPC1295-25GEA 915 EUR
APOSPC1295-25G APOSPC1295-5G
Perfluoro-2,5,8,11,14,17,20-heptamethyl-3,6,9,12,15,18,21-heptaoxatetracosanoyl fluoride 90%
Perfluoro-2,5,8,11,14,17,20-heptamethyl-3,6,9,12,15,18,21-heptaoxatetracosanoyl fluoride
Formula: C₂₄F₄₈O₈
Boiling Pt: 120…122 °C/1mmHg
Density: 1.8 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD03411205
CAS Number: 13140-25-5
UN: 3265
ADR: 8,II