3-Bromo-α,α,α-trifluorotoluene 98%
Supplier: Apollo Scientific
Warning
PC1340-1KG PC1340-250G PC1340-500G
APOSPC1340-1KGEA 197 EUR
APOSPC1340-1KG APOSPC1340-250G APOSPC1340-500G
3-Bromo-α,α,α-trifluorotoluene 98%
3-Bromo-α,α,α-trifluorotoluene
Formula: C₇H₄BrF₃
Boiling Pt: 154…156 °C
Density: 1.613 g/cm³ (25 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00000380
CAS Number: 401-78-5
EINECS: 206-932-6
UN: 1993
ADR: 3,III